Learn More
Galena, naturally occurring mineral, grains, approximately 0.06-0.19in, Thermo Scientific Chemicals
Description
Galena is used in infrared detectors, photo optic applications, slip property modifier used in friction industry for enhancing heat conduction and regulating friction coefficient, used as a semiconductor. Used in electronic devices and Infrared sensor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Black |
Physical Form | Powder, Flake, Crystalline, Beads, Lump, Etc. |
Quantity | 5 g |
Solubility Information | Soluble in water (0.00086g/L) and acid. Insoluble in alcohol,and potassium hydroxide. |
Formula Weight | 239.25 |
Odor | Odorless |
Chemical Name or Material | Galena, naturally occurring mineral, grains, approximately 0.06-0.19 in. |
Safety and Handling
Hazard Category | H302+H332-H350-H360Df-H373 |
Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c |
DOTInformation | Hazard Class: 9; Packaging Group: III |
EINECSNumber | 215-246-6 |
RTECSNumber | OG4550000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.