Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gallic Acid Monohydrate (Powder/Certified) ACS, Fisher Chemical

Click to view available options
Quantity:
500g
Packaging:
Poly Bottle
5995-86-8
C7H8O6
188.135
MFCD00002510
IUTKPPDDLYYMBE-UHFFFAOYSA-N
gallic acid monohydrate,3,4,5-trihydroxybenzoic acid hydrate,galop hydrate,3,4,5-trihydroxybenzoic acid monohydrate,gallicum acidum,benzoic acid, 3,4,5-trihydroxy-, hydrate,gallic acid, monohydrate,gallicacidmonohydrate,gallic acid, acs,acmc-1ary0
24721416
3,4,5-trihydroxybenzoic acid;hydrate
C1=C(C=C(C(=C1O)O)O)C(=O)O.O
5995-86-8
C7H8O6
188.135
MFCD00002510
IUTKPPDDLYYMBE-UHFFFAOYSA-N
gallic acid monohydrate,3,4,5-trihydroxybenzoic acid hydrate,galop hydrate,3,4,5-trihydroxybenzoic acid monohydrate,gallicum acidum,benzoic acid, 3,4,5-trihydroxy-, hydrate,gallic acid, monohydrate,gallicacidmonohydrate,gallic acid, acs,acmc-1ary0
24721416
3,4,5-trihydroxybenzoic acid;hydrate
C1=C(C=C(C(=C1O)O)O)C(=O)O.O
Additional Information Vapor Pressure: Negligible
Linear Formula C6H2(OH)3COOH·H2O
Density 1.69g/mL
Formula Weight 188.14
Physical Form Solid
Grade Certified
Identification Pass Test
Ignition Residue 0.05% max.
Packaging Poly Bottle
Solubility in Water Pass Test
Sulfate 0.02% max.
Water 10% max.
Decomposition Information 250°C
Melting Point 250°C
Color White
Quantity 500g
Chemical Name or Material Gallic Acid Monohydrate
Show More Show Less

WARNING!

Emergency Overview
Irritating to eyes, respiratory system and skin. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:2
Flammability:0
Instability:1