Gallium(III) chloride, ultra dry, 99.999% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 3569814

Quantity 25g

 View more versions of this product

Catalog No. AA3569814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13450-90-3
    Physical Form Ultra Dry. Various Forms (Powder/Flake/Crystalline/Beads, Etc.)
    Assay Percent Range 99.999% (Metal basis)
    Molecular Formula GaCl3
    Formula Weight 176.03
    MDL Number MFCD00011018
    Chemical Name or Material Gallium(III) chloride

    Specifications

    Physical Form Ultra Dry. Various Forms (Powder/Flake/Crystalline/Beads, Etc.)
    Formula Weight 176.03
    Quantity 25g
    Color White
    DOT Information DOT Class: 8, Packing Group: II
    Density 2.47g/cm3
    EINECS Number 236-610-0
    Assay Percent Range 99.999% (Metal basis)
    Molecular Formula GaCl3
    Appearance Ultra dry. White powder/flake/crystalline/beads, etc.
    MDL Number MFCD00011018
    CAS 13450-90-3
    Health Hazard 1 H301-H311-H332-H314-H318: Toxic if swallowed. Toxic in contact with skin. Harmful if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage.
    Chemical Name or Material Gallium(III) chloride

    H301-H311-H332-H314-H318: Toxic if swallowed. Toxic in contact with skin. Harmful if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit