Alfa Aesar™ Gallium(III) ethoxide

Manufacturer: Alfa Aesar™ 4485914

Quantity 25g

 View more versions of this product

Catalog No. AA4485914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2572-25-0
    Molecular Formula C6H15GaO3·xHOCH2CH3
    Linear Formula Ga(OC2H5)3·xHOCH2CH3
    Formula Weight 204.9
    MDL Number MFCD01866730
    Synonym Gallium triethoxide
    Chemical Name or Material Gallium (III) ethoxide

    Specifications

    Formula Weight 204.9
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338-P337+P313
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.
    CAS 2572-25-0
    Density 1.25g/mL
    Linear Formula Ga(OC2H5)3·xHOCH2CH3
    MDL Number MFCD01866730
    Color Yellow
    Sensitivity Moisture Sensitive
    Quantity 25g
    Physical Form Liquid
    Molecular Formula C6H15GaO3·xHOCH2CH3
    Synonym Gallium triethoxide
    Health Hazard 2 GHS H Statement
    H315-H319-H227
    Causes skin irritation.
    Causes serious eye irritation.
    Combustible liquid.
    Flash Point 68°C (154°F)
    Health Hazard 1 Warning
    Chemical Name or Material Gallium (III) ethoxide

    GHS H Statement
    H315-H319-H227
    Causes skin irritation.
    Causes serious eye irritation.
    Combustible liquid.

    GHS P Statement
    P280g-P305+P351+P338-P337+P313
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit