Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Gallium(III) ethoxide

Manufacturer:  Alfa Aesar™ 4485903

Quantity 1g

 View more versions of this product

Catalog No. AA4485903


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

2572-25-0
Ga(OC2H5)3·xHOCH2CH3
MFCD01866730
Gallium (III) ethoxide
C6H15GaO3·xHOCH2CH3
204.9
Gallium triethoxide

Specifications

Moisture Sensitive
2572-25-0
1.25g/mL
Ga(OC2H5)3·xHOCH2CH3
MFCD01866730
Yellow
Gallium (III) ethoxide
1g
Liquid
C6H15GaO3·xHOCH2CH3
204.9
Gallium triethoxide
68°C (154°F)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

GHS P Statement
P280g-P305+P351+P338-P337+P313
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit