Alfa Aesar™ Gallium(III) fluoride, anhydrous, 99.85% (metals basis)

Manufacturer: Alfa Aesar™ 3211203

Quantity 1g

 View more versions of this product

Catalog No. AA3211203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7783-51-9
    Formula Weight 126.72
    MDL Number MFCD00011021
    UN Number UN3260
    Merck Index 14,4354
    Chemical Name or Material Gallium(III) fluoride, Anhydrous

    Specifications

    Formula Weight 126.72
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Sensitivity Moisture Sensitive
    UN Number UN3260
    Boiling Point ≈1000°C
    Color White
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    CAS 7783-51-9
    Melting Point 800°C (subl.)
    Quantity 1g
    Health Hazard 1 Danger
    Density 4.47g/mL
    TSCA TSCA
    Physical Form Powder
    MDL Number MFCD00011021
    Merck Index 14,4354
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Chemical Name or Material Gallium(III) fluoride, Anhydrous
    Assay 99.85%, (metals basis)

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit