Alfa Aesar™ Gallium(III) iodide, ultra dry, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 4714806

Quantity 5g

 View more versions of this product

Catalog No. AA4714806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13450-91-4
    Formula Weight 450.43
    MDL Number MFCD00016107
    UN Number UN3260
    Chemical Name or Material Gallium(III) iodide, Ultra dry

    Specifications

    Formula Weight 450.43
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P260-P285-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    In case of inadequate ventilation wear respiratory protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Mesh Size −10 Mesh
    Health Hazard 2 GHS H Statement
    H334-H314-H318-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause an allergic skin reaction.
    DOT Information Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    Physical Form Beads
    Sensitivity Moisture & Light Sensitive
    MDL Number MFCD00016107
    CAS 13450-91-4
    UN Number UN3260
    Boiling Point 345°C (subl.)
    Color Yellow-green
    Melting Point 212°C
    Health Hazard 1 Danger
    Density 4.15g/mL
    TSCA TSCA
    Quantity 5g
    Storage Note 1
    • Ampouled under argon
    Chemical Name or Material Gallium(III) iodide, Ultra dry
    Assay 99.999%, (metals basis)

    GHS H Statement
    H334-H314-H318-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P285-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    In case of inadequate ventilation wear respiratory protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Ampouled under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit