Gallium (III) Nitrate Hydrate, 99.9%, metals basis, Alfa Aesar

Manufacturer: Alfa Aesar 3211602

Quantity 0.5g

 View more versions of this product

Catalog No. AA3211602

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 69365-72-6
    Physical Form Powder/Flake/Crystalline/Beads/Lump, Etc.
    Assay Percent Range 99.9% (Metal basis)
    Molecular Formula GaN3O9·xH2O
    Formula Weight 255.73 (anhydrous)
    MDL Number MFCD00149723
    Chemical Name or Material Gallium(III) nitrate hydrate

    Specifications

    Physical Form Powder/Flake/Crystalline/Beads/Lump, Etc.
    Formula Weight 255.73 (anhydrous)
    Quantity 0.5g
    Color White
    DOT Information DOT Class: 5.1, Packing Group: III
    EINECS Number 236-815-5
    Odor Odorless
    Assay Percent Range 99.9% (Metal basis)
    Molecular Formula GaN3O9·xH2O
    Appearance White powder/flake/crystalline/beads/lump, etc.
    MDL Number MFCD00149723
    CAS 69365-72-6
    Health Hazard 1 H272-H315-H319-H303: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May be harmful if swallowed.
    Chemical Name or Material Gallium(III) nitrate hydrate

    H272-H315-H319-H303: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May be harmful if swallowed.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit