Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gallium(III) nitrate hydrate, 99.9998%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
100g
Packaging:
Glass bottle
69365-72-6
GaN3O9·xH2O
255.74
MFCD00149723
YVFORYDECCQDAW-UHFFFAOYSA-N
gallium iii nitrate hydrate, nitric acid, gallium salt, hydrate, ga.3no3.h2o, gallium nitrate nanoparticles, gallium-iii-nitrate hydrate, gallium iii nitrate hydrate, puratronic, gallium iii nitrate hydrate, puratronic metals basis, gallium iii nitrate hydrate, crystalline trace metals basis, gallium iii nitrate hydrate, crystals and lumps trace metals basis
11550823
gallium;trinitrate;hydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[Ga+3]
69365-72-6
GaN3O9·xH2O
255.74
MFCD00149723
YVFORYDECCQDAW-UHFFFAOYSA-N
gallium iii nitrate hydrate, nitric acid, gallium salt, hydrate, ga.3no3.h2o, gallium nitrate nanoparticles, gallium-iii-nitrate hydrate, gallium iii nitrate hydrate, puratronic, gallium iii nitrate hydrate, puratronic metals basis, gallium iii nitrate hydrate, crystalline trace metals basis, gallium iii nitrate hydrate, crystals and lumps trace metals basis
11550823
gallium;trinitrate;hydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[Ga+3]

Gallium(III) nitrate hydrate, 99.9998%, (trace metal basis), ACROS Organics, Quantity: 100g, Packaging: Glass bottle, CAS: 69365-72-6, White, Grade: Trace Metal Basis, Melting Point: 110.0 deg.C, Molecular Formula: GaN3O9.xH2O, Molecular Weight: 255.74, Percent Purity: 99.9998%

Gallium(III) nitrate hydrate, 99.9998%, (trace metal basis), ACROS Organics, Quantity: 100g, Packaging: Glass bottle, CAS: 69365-72-6, White, Grade: Trace Metal Basis, Melting Point: 110.0 deg.C, Molecular Formula: GaN3O9.xH2O, Molecular Weight: 255.74, Percent Purity: 99.9998%

for ultra trace analysis
Assay Percent Range 99.9998%
Linear Formula Ga(NO3)3·xH2O
Merck Index 14, 4350
Solubility Information Solubility in water: soluble
Formula Weight 255.74
Physical Form Crystalline Powder, Crystals, and/or Chunks
Percent Purity 99.9998%
Grade Trace Metal Basis
Packaging Glass bottle
Solubility Solubility in water: soluble.
Total Trace Metal Impurities 0.0002% max
Color White
Melting Point 110.0°C
Quantity 5g
Chemical Name or Material Gallium(III) nitrate hydrate
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May intensify fire; oxidizer.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger