Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gallium(III) nitrate solution, Ga 9-10% w/w, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
Gallium(III) nitrate solution, is used as the material for producing gallium salts and for scientific research and chemical reagent. It is also used as an intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Liquid |
Quantity | 250 g |
UN Number | UN3218 |
Solubility Information | It is soluble in water. |
Formula Weight | 255.73 |
Concentration or Composition (by Analyte or Components) | Ga 9 to 10% w/w |
Chemical Name or Material | Gallium (III) nitrate solution |
Safety and Handling
Hazard Category | H271-H290-H314-H331 |
Hazard Statement | GHS H Statement H272-H314-H318 May intensify fire oxidizer. Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P210-P220-P221-P234-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c |
DOTInformation | Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, AQUEOUS SOLUTION, N.O.S . |
EINECSNumber | 236-815-5 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction