Alfa Aesar™ Gallium(III) nitride, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 4021830

Quantity 250g

 View more versions of this product

Catalog No. AA4021830

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 25617-97-4
    Formula Weight 83.73
    MDL Number MFCD00016108
    Merck Index 14,4351
    Chemical Name or Material Gallium(III) nitride

    Specifications

    Formula Weight 83.73
    Health Hazard 3 GHS P Statement
    P261-P280-P363-P321-P333+P313-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    If skin irritation or rash occurs: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Solubility Information Insoluble in water, dilute acids; Slightly soluble in hot conc; H2SO4, hot conc; NaOH
    Physical Form Powder
    CAS 25617-97-4
    Merck Index 14,4351
    Health Hazard 1 Warning
    Density 6.1g/mL
    TSCA TSCA
    Sensitivity Moisture Sensitive
    MDL Number MFCD00016108
    Color Yellow
    Melting Point 800°C (subl.)
    Quantity 250g
    Health Hazard 2 GHS H Statement
    H317
    May cause an allergic skin reaction.
    Chemical Name or Material Gallium(III) nitride
    Assay 99.99%, (metals basis)

    GHS H Statement
    H317
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P363-P321-P333+P313-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    If skin irritation or rash occurs: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit