Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gallium(III) oxide, 99.999% (metals basis), Alfa Aesar

For Use As An Evaporated Material and Sputtering Target of 99.999%, in Dielectric Films

Manufacturer:  Alfa Aesar 1050814

Quantity 25g

 View more versions of this product

Catalog No. AA1050814


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12024-21-4
99.999% (Metal basis)
187.44
Gallium(III) oxide
Powder or Lump
Ga2O3
MFCD00011020

Specifications

12024-21-4
99.999% (Metal basis)
White powder or lump
MFCD00011020
White
Gallium(III) oxide
Powder or Lump
Ga2O3
187.44
Odorless
25g

Safety and Handling

SDS & Safety Documents

EINECSNumber : 234-691-7

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit