Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gallium(III) oxide, Puratronic™, 99.999% (metals basis), Alfa Aesar™

Click to view available options
Quantity:
5g
25g
100g
500g
12024-21-4
Ga2O3
MFCD00011020
12024-21-4
Ga2O3
MFCD00011020

Used in vacuum deposition.

Density 6.44
Refractive Index 1.92
Packaging Plastic bottle
Specific Gravity 6.44
Quantity 25g
Melting Point 1900°C
Solubility Information Soluble in acids. Insoluble in water
Formula Weight 187.44
Physical Form -50 Mesh Powder
Percent Purity 99.999%
Chemical Name or Material Gallium(III) oxide
Show More Show Less

EINECSNumber : 234-691-7

TSCA : Yes

Recommended Storage : Ambient temperatures