Gallium(III) perchlorate hydrate, Crystalline, 13.5-15.5% Ga, Alfa Aesar™

Manufacturer: Alfa Aesar 45580-04

Quantity 2g

 View more versions of this product

Catalog No. AA4558004

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 81029-07-4
    Physical Form Crystalline
    Assay Percent Range 13.5-15.5% Ga
    Molecular Formula Cl3GaO12·xH2O
    Formula Weight 368.07 (Anhydrous)
    MDL Number MFCD00167156
    Chemical Name or Material Gallium(III) perchlorate hydrate

    Specifications

    Formula Weight 368.07 (Anhydrous)
    Quantity 2g
    DOT Information DOT Class: 5.1, Packing Group: II
    Physical Form Crystalline
    Assay Percent Range 13.5-15.5% Ga
    Molecular Formula Cl3GaO12·xH2O
    MDL Number MFCD00167156
    CAS 81029-07-4
    Health Hazard 1 H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Gallium(III) perchlorate hydrate

    H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit