Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gallium Ingot, 99.9999% (Metals basis), Alfa Aesar™

Requires overnight shipping with cold pack

Manufacturer:  Alfa Aesar 1018536

Quantity 500g

 View more versions of this product

Catalog No. AA1018536


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7440-55-3
99.9999% (Metals basis)
Gallium
Ingot
MFCD00134045

Specifications

500g
Silver Gray ingot
99.9999% (Metals basis)
Silver-gray
7440-55-3
Ingot
MFCD00134045
Gallium

Safety and Handling

SDS & Safety Documents

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 231-163-8

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit