Alfa Aesar™ Gallium pellets, 6mm dia, 99.99999% (metals basis)

Manufacturer: Alfa Aesar™ 4432518

Quantity 50g

 View more versions of this product

Catalog No. AA4432518

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-55-3
    MDL Number MFCD00134045
    UN Number UN2803
    Chemical Name or Material Gallium pellets, 6mm dia.

    Specifications

    Physical Form Pellet
    Color Silver gray
    Quantity 50g
    UN Number UN2803
    Health Hazard 1 Warning
    Recommended Storage 2°C to 8°C, Requires overnight shipping with cold pack
    RTECS Number LW8600000
    Weight Max. weight 0.7g/pellet
    TSCA TSCA
    MDL Number MFCD00134045
    CAS 7440-55-3
    Health Hazard 3 GHS P Statement
    P234-P390-P406
    Keep only in original container.
    Absorb spillage to prevent material damage.
    Store in corrosive resistant container with a resistant inner liner.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: GALLIUM
    Health Hazard 2 GHS H Statement
    H290
    May be corrosive to metals.
    Chemical Name or Material Gallium pellets, 6mm dia.
    Assay 99.99999%, (metals basis)

    GHS H Statement
    H290
    May be corrosive to metals.

    2°C to 8°C, Requires overnight shipping with cold pack

    GHS P Statement
    P234-P390-P406
    Keep only in original container.
    Absorb spillage to prevent material damage.
    Store in corrosive resistant container with a resistant inner liner.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit