Alfa Aesar™ Gallium Tin alloy, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 4365509

Quantity 10g

 View more versions of this product

Catalog No. AA4365509

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00144390
    UN Number UN2759
    Chemical Name or Material Gallium Tin alloy

    Specifications

    Physical Form Low Melting Solid
    TSCA TSCA
    Odor Odorless
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H290
    May be corrosive to metals.
    Health Hazard 3 GHS P Statement
    P234-P390-P406
    Keep only in original container.
    Absorb spillage to prevent material damage.
    Store in corrosive resistant container with a resistant inner liner.
    Quantity 10g
    MDL Number MFCD00144390
    UN Number UN2759
    Color Silver
    Melting Point ≥28°C
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
    Avg. Mol. Wt. or Mol. Wt. Range Ga:Sn; 98:2 wt%
    Chemical Name or Material Gallium Tin alloy
    Assay 99.99%, (metals basis)

    GHS H Statement
    H290
    May be corrosive to metals.

    GHS P Statement
    P234-P390-P406
    Keep only in original container.
    Absorb spillage to prevent material damage.
    Store in corrosive resistant container with a resistant inner liner.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit