Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Gallium Tin alloy, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 4365518

Quantity 50g

 View more versions of this product

Catalog No. AA4365518


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

MFCD00144390
UN2759
Gallium Tin alloy

Specifications

Ga:Sn; 98:2 wt%
Low Melting Solid
UN2759
Silver
Gallium Tin alloy
50g
MFCD00144390
Odorless
≥28°C
99.99%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H290
May be corrosive to metals.

GHS P Statement
P234-P390-P406
Keep only in original container.
Absorb spillage to prevent material damage.
Store in corrosive resistant container with a resistant inner liner.

Warning

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit