Gallium Tin alloy, 99.99% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1816118

Quantity 50g

 View more versions of this product

Catalog No. AA1816118

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-31-5,7440-55-3
    Physical Form Liquid
    Assay Percent Range 99.99% (metals basis)
    Molecular Formula Ga:Sn; 92:8 wt.%
    MDL Number MFCD00144390
    Chemical Name or Material Gallium Tin alloy

    Specifications

    Physical Form Liquid
    Quantity 50g
    Color Silver
    DOT Information Hazard Class: 8, Packing Group: III
    Health Hazard 2 Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
    Assay Percent Range 99.99% (metals basis)
    CAS 7440-55-3,7440-31-5
    Molecular Formula Ga:Sn; 92:8 wt.%
    MDL Number MFCD00144390
    Appearance Silver-colored liquid
    Concentration or Composition (by Analyte or Components) Tin: 8.0%; Gallium: 92.0%
    Health Hazard 1 H290: May be corrosive to metals.
    Chemical Name or Material Gallium Tin alloy

    Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

    H290: May be corrosive to metals.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit