Learn More
Sklar InstrumentsSupplier Diversity Partner Luer-Koerte Gallstone Scoops, Sklar, 12-3/4 in., 3.2mm, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 321051
Luer-Koerte Gallstone Scoops, Sklar, 12-3/4 in., 3.2mm, Stainless Steel. Sklar's Luer-Koert Gall Stone Scoops allows for easy probing and removal of gall stones in tubular ducts. The scoops are available in various sizes. This product has a malleable 12-3/4 inch shaft with a 3.2mm scoop.
Catalog No. 50-143-8182