Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GalNAc beta(1-3)Gal alpha(1-4)Gal beta(1-4)Glc-beta-pNP 88.0+%, TCI America™

Click to view available options
Quantity:
5mg
1134635-03-2
C32H48N2O23
828.727
AUMWVXPGKMCSQE-CUWUVIFCSA-N
53384359
N-[(2S,3S,4R,5R,6S)-2-[(2R,3S,4S,5S,6S)-2-[(2S,3R,4R,5S,6S)-6-[(2S,3S,4R,5S,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy-4,5-dihydroxy-6-(hydr
CC(=O)NC1C(C(C(OC1OC2C(C(OC(C2O)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5=CC=C(C=C5)[N+](=O)[O-])CO)CO)CO)O)CO)O)O
1134635-03-2
C32H48N2O23
828.727
AUMWVXPGKMCSQE-CUWUVIFCSA-N
53384359
N-[(2S,3S,4R,5R,6S)-2-[(2R,3S,4S,5S,6S)-2-[(2S,3R,4R,5S,6S)-6-[(2S,3S,4R,5S,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy-4,5-dihydroxy-6-(hydr
CC(=O)NC1C(C(C(OC1OC2C(C(OC(C2O)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5=CC=C(C=C5)[N+](=O)[O-])CO)CO)CO)O)CO)O)O
Color White-Yellow
Quantity 5mg
Formula Weight 828.73
Physical Form Crystal-Powder at 20°C
Percent Purity ≥88.0% (HPLC)
Chemical Name or Material GalNAc beta(1-3)Gal alpha(1-4)Gal beta(1-4)Glc-beta-pNP

TSCA : No

Recommended Storage : Freezer