Learn More
Aviva Systems Biology Corp GALNT10 Antibody - N-terminal region (ARP48800_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP48800_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GALNT10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GALNT10. Alternate names: DKFZp586H0623, FLJ00205, FLJ11715, GalNAcT10, pp-GalNAc-T10, GALNACT10, PPGALNACT10, PPGANTASE10ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59926