Learn More
Aviva Systems Biology Corp GALNT5 Antibody - middle region (ARP47040_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47040_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GALNT5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GALNT5. Alternate names: GALNAC-T5, MGC165041ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59456