Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GALNT6 Recombinant Rabbit Monoclonal Antibody (HL2392)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA550516
Description
GALNT6 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
This gene encodes a member of the UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAc-T) family of enzymes. GalNAc-Ts initiate mucin-type O-linked glycosylation in the Golgi apparatus by catalyzing the transfer of GalNAc to serine and threonine residues on target proteins. They are characterized by an N-terminal transmembrane domain, a stem region, a lumenal catalytic domain containing a GT1 motif and Gal/GalNAc transferase motif, and a C-terminal ricin/lectin-like domain. GalNAc-Ts have different, but overlapping, substrate specificities and patterns of expression. The encoded protein is capable of glycosylating fibronectin peptide in vitro and is expressed in a fibroblast cell line, indicating that it may be involved in the synthesis of oncofetal fibronectin.
Specifications
GALNT6 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q8C7U7, Q8NCL4 | |
GALNT6 | |
Recombinant fragment of human GALNT6. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
HL2392 | |
Unconjugated | |
GALNT6 | |
4632410F13; AW047994; fc23d02; fc56f12; GalNAc transferase 6; GalNAcT6; galNAc-T6; GALNT6; N-acetylgalactosaminyltransferase T-6; polypeptide GalNAc transferase 6; Polypeptide N-acetylgalactosaminyltransferase 6; pp-GaNTase; pp-GaNTase 6; protein-UDP acetylgalactosaminyltransferase 6; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 6; UDP-N-acetyl-alpha-D-galactosamine: polypeptide N-acetylgalactosaminyltransferase 6; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 6; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 6 (GalNAc-T6); wu:fc23d02; wu:fc56f12; zgc:77836 | |
Rabbit | |
Protein A | |
RUO | |
11226, 207839 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction