Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galvinoxyl free radical, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
2370-18-5
C29H41O2
421.64
MFCD00001616
2,6-Di-tert-butyl-α-(3,5-di-tert-butyl-4-oxo-2,5-cyclohexadien-1-ylidene)-p-tolyloxy free radical
2370-18-5
C29H41O2
421.64
MFCD00001616
2,6-Di-tert-butyl-α-(3,5-di-tert-butyl-4-oxo-2,5-cyclohexadien-1-ylidene)-p-tolyloxy free radical
Polymerization reagent
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Blue to Purple
Melting Point 156°C to 160°C
Assay Percent Range 98%
Beilstein 08, IV, 1379
Fieser 01,409
Merck Index 15, 4387
Formula Weight 421.64
Physical Form Crystalline Powder, Crystals and/or Chunks
Percent Purity ≥97.5%
Chemical Name or Material Galvinoxyl free radical
Show More Show Less