Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galvinoxyl free radical, 98%, Thermo Scientific Chemicals

Catalog Number AC119850250
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
2370-18-5
C29H42O2
422.65
MFCD00001616
HRLDHROCDBYHAU-UHFFFAOYSA-N
2,6-Di-tert-butyl-α-(3,5-di-tert-butyl-4-oxo-2,5-cyclohexadien-1-ylidene)-p-tolyloxy free radical
2,6-di-tert-butyl-4-[(3,5-di-tert-butyl-4-hydroxyphenyl)methylidene]cyclohexa-2,5-dien-1-one
CC(C)(C)C1=CC(C=C2C=C(C(=O)C(=C2)C(C)(C)C)C(C)(C)C)=CC(=C1O)C(C)(C)C
2370-18-5
C29H42O2
422.65
MFCD00001616
HRLDHROCDBYHAU-UHFFFAOYSA-N
2,6-Di-tert-butyl-α-(3,5-di-tert-butyl-4-oxo-2,5-cyclohexadien-1-ylidene)-p-tolyloxy free radical
2,6-di-tert-butyl-4-[(3,5-di-tert-butyl-4-hydroxyphenyl)methylidene]cyclohexa-2,5-dien-1-one
CC(C)(C)C1=CC(C=C2C=C(C(=O)C(=C2)C(C)(C)C)C(C)(C)C)=CC(=C1O)C(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Polymerization reagent
TRUSTED_SUSTAINABILITY
Melting Point 156°C to 160°C
Color Blue to Purple
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 25 g
Beilstein 08, IV, 1379
Fieser 01,409
Merck Index 15, 4387
Formula Weight 421.64
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Galvinoxyl free radical
Show More Show Less
EINECSNumber 219-133-2

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit