Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gamma Adaptin Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1107877
Description
Gamma Adaptin Polyclonal specifically detects Gamma Adaptin in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
Gamma Adaptin | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
Adapter-related protein complex 1 subunit gamma-1, Adaptor protein complex AP-1 subunit gamma-1, adaptor-related protein complex 1, gamma 1 subunit, ADTGclathrin assembly protein complex 1 gamma large chain, AP-1 complex subunit gamma-1, CLAPG1clathrin-associated/assembly/adaptor protein, large, gamma 1, Clathrin assembly protein complex 1 gamma-1 large chain, gamma adaptin, gamma1-adaptin, golgi adaptor HA1/AP1 adaptin gamma subunit, Golgi adaptor HA1/AP1 adaptin subunit gamma-1, MGC18255 | |
Goat | |
91.35 kDa | |
0.1 mg | |
Golgi Apparatus Markers, Membrane Trafficking and Chaperones | |
164 | |
Human, Orangutan, Primate | |
IgG |
Western Blot, ELISA, Immunoprecipitation | |
0.7 mg/ml | |
Western Blot 1:500-1:2000, ELISA 1:2000-1:8000, Immunoprecipitation 1:100 | |
O43747 | |
AP1G1 | |
This affinity purified Gamma Adaptin Antibody was prepared from whole goat serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near aa 625-675 of Human Gamma Adaptin. (Uniprot: O43747) | |
Affinity Purified | |
RUO | |
Primary | |
AP1G1 | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction