Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ gamma Adaptin Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01670559
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-559 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-559 Supplier Invitrogen™ Supplier No. PA118306
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent.

Adaptins are important components of clathrin-coated vesicles transporting ligand-receptor complexes from the plasma membrane or from the trans-Golgi network to lysosomes. The adaptin family of proteins is composed of four classes of molecules named alpha, beta-, beta prime- and gamma- adaptins. Adaptins, together with medium and small subunits, form a heterotetrameric complex called an adaptor, whose role is to promote the formation of clathrin-coated pits and vesicles. The protein encoded by this gene is a gamma-adaptin protein and it belongs to the adaptor complexes large subunits family. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen gamma Adaptin
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 0.02% thimerosal
Gene AP1G1
Gene Accession No. O43747
Gene Alias AA409002; adapter-related protein complex 1 subunit gamma-1; adaptor protein complex AP-1 gamma 1 subunit; adaptor protein complex AP-1 subunit gamma-1; adaptor protein complex AP-1, gamma 1 subunit; adaptor related protein complex 1 gamma 1 subunit; adaptor related protein complex 1 subunit gamma 1; adaptor-related protein complex 1 gamma 1 subunit; adaptor-related protein complex 1 subunit gamma-1; adaptor-related protein complex 1, gamma 1 subunit; ADTG; AP-1 complex subunit gamma-1; Ap1g1; AU041323; AW551707; CLAPG1; clathrin assembly protein complex 1 gamma large chain; clathrin assembly protein complex 1 gamma-1 large chain; clathrin-associated/assembly/adaptor protein, large, gamma 1; D8Ertd374e; gamma adaptin; Gamma1-adaptin; gamma-adaptin; golgi adaptor HA1/AP1 adaptin gamma subunit; golgi adaptor HA1/AP1 adaptin subunit gamma-1; MGC18255; testicular tissue protein Li 21
Gene Symbols AP1G1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the N-terminal residues I(5) R L R E L I R T I R T A R T Q A E E R(24) of human AP-1 complex subunit gamma-1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 164, 171494
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.