Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gamma Adaptin Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | Gamma Adaptin |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP157633
|
Novus Biologicals
NBP157633 |
100 μL |
Each of 1 for $436.00
|
|
Description
Gamma Adaptin Polyclonal specifically detects Gamma Adaptin in Human samples. It is validated for Western Blot.Specifications
Gamma Adaptin | |
Polyclonal | |
Rabbit | |
Golgi Apparatus Markers, Membrane Trafficking and Chaperones | |
O43747 | |
164 | |
Synthetic peptides corresponding to AP1G1(adaptor-related protein complex 1, gamma 1 subunit) The peptide sequence was selected from the C terminal of AP1G1. Peptide sequence DHMRSALLERMPVMEKVTTNGPTEIVQTNGETEPAPLETKPPPSGPQPTS. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
Adapter-related protein complex 1 subunit gamma-1, Adaptor protein complex AP-1 subunit gamma-1, adaptor-related protein complex 1, gamma 1 subunit, ADTGclathrin assembly protein complex 1 gamma large chain, AP-1 complex subunit gamma-1, CLAPG1clathrin-associated/assembly/adaptor protein, large, gamma 1, Clathrin assembly protein complex 1 gamma-1 large chain, gamma adaptin, gamma1-adaptin, golgi adaptor HA1/AP1 adaptin gamma subunit, Golgi adaptor HA1/AP1 adaptin subunit gamma-1, MGC18255 | |
AP1G1 | |
IgG | |
Affinity Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title