Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
gamma Catenin Antibody ([11E4) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254528200UG
Description
gamma Catenin Monoclonal antibody specifically detects gamma Catenin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.Specifications
gamma Catenin | |
Monoclonal | |
1 mg/mL | |
Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, CyTOF-ready | |
catenin (cadherin-associated protein), gamma (80kD), catenin (cadherin-associated protein), gamma 80kDa, Catenin gamma, Desmoplakin III, Desmoplakin-3, DP3CTNNG, DPIII, gamma-catenin, junction plakoglobin, PDGBARVD12, PKGB | |
Mouse | |
84 kDa | |
200 μg | |
Cancer, Cytoskeleton Markers | |
3728 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF | |
11 E4 | |
Unconjugated | |
PBS with No Preservative | |
JUP | |
Recombinant full length human plakoglobin, fused to maltose binding protein | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction