Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
gamma Catenin Mouse anti-Human, Mouse, Clone: 11 E4, Novus Biologicals™
Mouse Monoclonal Antibody
$240.50 - $482.00
Specifications
Antigen | gamma Catenin |
---|---|
Clone | 11 E4 |
Concentration | 0.2 mg/mL |
Dilution | Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25330620
|
Novus Biologicals
NBP25330620UG |
20 μg |
Each for $240.50
|
|
|||||
NB253306100
|
Novus Biologicals
NBP253306100UG |
100 μg |
Each for $482.00
|
|
|||||
NB253306200
|
Novus Biologicals
NBP253306200UG |
200 μg | Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. | N/A | |||||
Description
gamma Catenin Monoclonal specifically detects gamma Catenin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
gamma Catenin | |
0.2 mg/mL | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Cancer, Cytoskeleton Markers, Phospho Specific | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
3728 | |
Recombinant full length human plakoglobin, fused to maltose binding protein | |
Primary | |
Store at 4C. |
11 E4 | |
Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
catenin (cadherin-associated protein), gamma (80kD), catenin (cadherin-associated protein), gamma 80kDa, Catenin gamma, Desmoplakin III, Desmoplakin-3, DP3CTNNG, DPIII, gamma-catenin, junction plakoglobin, PDGBARVD12, PKGB | |
JUP | |
IgG1 κ | |
Protein A or G purified | |
84 kDa |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title