Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
gamma Catenin Antibody ([11E4), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254528PEATT594
Description
gamma Catenin Monoclonal specifically detects gamma Catenin in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
gamma Catenin | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
JUP | |
Recombinant full length human plakoglobin, fused to maltose binding protein (Uniprot: P14923) | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG1 κ |
Flow Cytometry | |
[11E4] | |
Flow Cytometry | |
catenin (cadherin-associated protein), gamma (80kD), catenin (cadherin-associated protein), gamma 80kDa, Catenin gamma, Desmoplakin III, Desmoplakin-3, DP3CTNNG, DPIII, gamma-catenin, junction plakoglobin, PDGBARVD12, PKGB | |
Mouse | |
Protein A or G purified | |
Cancer, Cytoskeleton Markers, Phospho Specific | |
3728.0 | |
Store at 4°C in the dark. Do not freeze. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction