Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gamma Catenin Mouse anti-Human, Mouse, Rat, Bovine, Canine, Chicken, Feline, Alexa Fluor™ 750, Clone: 15F11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254527AF750

Catalog No. NP2427A70


Add to cart

Description

Description

gamma Catenin Monoclonal antibody specifically detects gamma Catenin in Human,Mouse,Rat,Bovine,Canine,Chicken,Feline samples. It is validated for Western Blot,Flow Cytometry,Immunocytochemistry,Immunofluorescence.
Specifications

Specifications

gamma Catenin
15F11
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
catenin (cadherin-associated protein), gamma (80kD), catenin (cadherin-associated protein), gamma 80kDa, Catenin gamma, Desmoplakin III, Desmoplakin-3, DP3CTNNG, DPIII, gamma-catenin, junction plakoglobin, PDGBARVD12, PKGB
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Bovine, Canine, Chicken, Feline, Human, Mouse, Rat
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
JUP
Recombinant, full length chicken Catenin gamma protein
0.1 mL
Primary
3728
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit