Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gamma Catenin Mouse anti-Human, PE/Cy5.5, Clone: CTNG/1483, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254525PECY55

Catalog No. N254525PEC5


Add to cart

Description

Description

gamma Catenin Monoclonal antibody specifically detects gamma Catenin in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

gamma Catenin
CTNG/1483
Flow Cytometry
Conjugated, Purified
JUP
Recombinant human full-length gamma catenin protein
Protein A or G purified
Cancer, Cytoskeleton Markers, Phospho Specific
Primary
3728.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
catenin (cadherin-associated protein), gamma (80kD), catenin (cadherin-associated protein), gamma 80kDa, Catenin gamma, Desmoplakin III, Desmoplakin-3, DP3CTNNG, DPIII, gamma-catenin, junction plakoglobin, PDGBARVD12, PKGB
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit