Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gamma Catenin Rabbit anti-Human, Rat, Bovine, Canine, Chicken, Feline, Clone: CTNG/2155R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen gamma Catenin
Clone CTNG/2155R
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122661
View Documents
Novus Biologicals
NBP30759410UG
100 μg
Each for $406.52
Add to cart
 
NB122662
View Documents
Novus Biologicals
NBP30759420UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

gamma Catenin Monoclonal antibody specifically detects gamma Catenin in Human, Rat, Bovine, Canine, Chicken, Feline samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Specifications

Specifications

gamma Catenin
0.2 mg/ml
Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Purified
RUO
Human, Rat, Bovine, Canine, Chicken, Feline
catenin (cadherin-associated protein), gamma (80kD), catenin (cadherin-associated protein), gamma 80kDa, Catenin gamma, Desmoplakin III, Desmoplakin-3, DP3CTNNG, DPIII, gamma-catenin, junction plakoglobin, PDGBARVD12, PKGB
Recombinant, human full-length gamma Catenin (CTNG) protein
Monoclonal
Protein A or G purified
CTNG/2155R
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Unconjugated
Rabbit
Cancer, Cell Biology, Cellular Markers, Phospho Specific
10 mM PBS with 0.05% BSA
3728
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit