Learn More
gamma-Glutamylcyclotransferase/CRF21/GGCT Rabbit anti-Human, Janelia Fluor 549, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP300283JF549
Catalog No. NB065307
Add to cart
Description
Description
gamma-Glutamylcyclotransferase/CRF21/GGCT Polyclonal antibody specifically detects gamma-Glutamylcyclotransferase/CRF21/GGCT in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
gamma-Glutamylcyclotransferase/CRF21/GGCT
Janelia Fluor 549
50mM Sodium Borate
gamma-glutamylcyclotransferase, GCTG, GGC, MGC3077
Produced in rabbits immunized with purified, recombinant gamma-Glutamylcyclotransferase/CRF21/GGCT (O75223-1; Met1-Leu188).
Affinity purified
RUO
Primary
79017
Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human