Learn More
Alfa Aesar™ gamma-Neuropeptide, Rabbit
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66216EXD
 View more versions of this product
Catalog No. AAJ66216EXD
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
2320.61
Rabbit
Solid
RUO
γ-Preprotachykinin (72-92), amide
HPLC
0.5mg
Asp-Ala-Gly-His-Gly-Gln-Ile-Ser-His-Lys-Arg-His-Lys-Thr-Asp-Ser-Phe-Val-Gly-Leu-Met-NH2
-30° to -10°C
gamma-Neuropeptide, rabbit