Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ gamma-Phenyl-gamma-butyrolactone, 97%

Manufacturer:  Alfa Aesar™ B2436406

Quantity 5g

 View more versions of this product

Catalog No. AAB2436406


  Check Availability
Add to cart

Specifications

Description

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

1008-76-0
162.19
125169
gamma-Phenyl-gamma-butyrolactone
C10H10O2
MFCD00005398
4,5-Dihydro-5-phenyl-2(3H)-furanone

Specifications

5g
C10H10O2
MFCD00005398
4,5-Dihydro-5-phenyl-2(3H)-furanone
35° to 38°C
gamma-Phenyl-gamma-butyrolactone
1008-76-0
162.19
125169
305°C
>110°C (230°F)
97%

Safety and Handling

SDS & Safety Documents

RTECSNumber : LU3690000

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit