Learn More
Alfa Aesar™ gamma-Secretase Inhibitor II
Manufacturer:  Alfa Aesar™ J64828MCR
 View more versions of this product
Catalog No. AAJ64828MCR
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1mg
-30°C to -10°C
gamma-Secretase Inhibitor
C20H30N2O4
MW167
Off-white
Solid
A cell-permeable, reversible and selective peptidomimetic inhibitor of --secretase
362.47