Learn More
Thermo Scientific™ gamma-Secretase Inhibitor II
Supplier: Thermo Scientific™ J64828LB0
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
5 mg | |
-30°C to -10°C | |
C20H30N2O4 | |
MW167 | |
Solid |
Off-white | |
A cell-permeable, reversible and selective peptidomimetic inhibitor of --secretase | |
362.47 | |
gamma-Secretase Inhibitor |