Learn More
Genetex IncSupplier Diversity Partner Anti-gamma Synuclein antibody DFS Item
Manufacturer:  Genetex IncSupplier Diversity Partner GTX110483
The Anti-gamma Synuclein antibody from GeneTex is a Rabbit Polyclonal antibody to gamma Synuclein. The Anti-gamma Synuclein antibody Recognizes Human, Mouse. The Anti-gamma Synuclein antibody has been validated for the following Applications: ICC/IF, IHC, IHC-P, WB.
Catalog No. NC1462771