Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gamma Tubulin Mouse anti-Human, Mouse, Rat, Drosophila, HRP, Clone: 4511, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243585H

Catalog No. NBP243585H


Add to cart

Description

Description

gamma Tubulin Monoclonal antibody specifically detects gamma Tubulin in Human, Mouse, Rat, Drosophila samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

gamma Tubulin
4511
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
TUBG1
Recombinant protein encompassing a sequence within the center region of human gamma Tubulin. The exact sequence is proprietary.
Protein G purified
Cell Biology, Cellular Markers, Centrosome Markers, Cytoskeleton Markers
Primary
7283
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
HRP
PBS with No Preservative
gamma-1-tubulin, Gamma-tubulin complex component 1, GCP-1, TUBG, TUBGCP1, tubulin gamma-1 chain, tubulin, gamma 1, tubulin, gamma polypeptide
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat, Drosophila
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit