Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ganaxolone, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
38398-32-2
C22H36O2
332.528
PGTVWKLGGCQMBR-DFTCEIPHSA-N
ganaxolone,3alpha-hydroxy-3beta-methyl-5alpha-pregnan-20-one,1-3as,3br,5as,7r,9as,9bs,11as-7-hydroxy-7,9a,11a-trimethyl-tetradecahydrocyclopenta a phenanthren-1-yl ethanone
92043431
1-[(3R,5S,8R,9S,10S,13S,14S)-3-hydroxy-3,10,13-trimethyl-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]ethanone
CC(=O)C1CCC2C1(CCC3C2CCC4C3(CCC(C4)(C)O)C)C
This item is not returnable. View return policy
38398-32-2
C22H36O2
332.528
PGTVWKLGGCQMBR-DFTCEIPHSA-N
ganaxolone,3alpha-hydroxy-3beta-methyl-5alpha-pregnan-20-one,1-3as,3br,5as,7r,9as,9bs,11as-7-hydroxy-7,9a,11a-trimethyl-tetradecahydrocyclopenta a phenanthren-1-yl ethanone
92043431
1-[(3R,5S,8R,9S,10S,13S,14S)-3-hydroxy-3,10,13-trimethyl-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]ethanone
CC(=O)C1CCC2C1(CCC3C2CCC4C3(CCC(C4)(C)O)C)C

Potent, positive allosteric modulator of GABAA receptors

Potent positive allosteric modulator of GABA A receptors. Enhances GABA-evoked chloride currents in Xenopus oocytes expressing GABA A receptors (EC 50 values are 94, 122 and 213 nM for α2β1γ2 L α3β1γ2 L and α1β1γ2 L receptors respectively). Exerts anticonvulsive effects in a broad range of animal seizure models.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 332.53
Percent Purity >98%
Chemical Name or Material Ganaxolone

Recommended Storage : Store at +4°C