Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ganciclovir, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
1 kg
Packaging:
Amber Glass Bottle

G2211, 82410-32-0, C9H13N5O4

Spectrum™ Ganciclovir, USP is an anti-viral drug. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Ignition Residue 0.001
Quantity 1 kg
Water 0.06
CAS 82410-32-0
CAS Max % 1
Molecular Formula C9H13N5O4
InChI Key IRSCQMHQWWYFCW-UHFFFAOYSA-N
SMILES NC1=NC(=O)C2=C(N1)N(COC(CO)CO)C=N2
IUPAC Name 2-amino-9-{[(1,3-dihydroxypropan-2-yl)oxy]methyl}-6,9-dihydro-3H-purin-6-one
Molecular Weight (g/mol) 255.23
Percent Purity 98 to 102%
Show More Show Less