Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gankyrin Mouse anti-Human, Clone: 1F4, Novus Biologicals™
Mouse Monoclonal Antibody
$337.00 - $482.50
Specifications
Antigen | Gankyrin |
---|---|
Clone | 1F4 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1012, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25940050
|
Novus Biologicals
NBP25940050UL |
50 μL |
Each for $337.00
|
|
|||||
NBP259400X
|
Novus Biologicals
NBP259400100UL |
100 μL |
Each for $482.50
|
|
|||||
Description
Gankyrin Monoclonal specifically detects Gankyrin in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Gankyrin | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Cancer, Cell Biology, Tumor Suppressors, Ubiquitin Proteasome Pathway | |
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
5716 | |
Recombinant human PSMD10 (1-226aa) purified from E. coli | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1F4 | |
Western Blot 1:1000, Flow Cytometry 1:10-1:1012, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
26S proteasome non-ATPase regulatory subunit 10, 26S proteasome regulatory subunit p28, ankyrin repeat protein, dJ889N15.2, gankyrin, hepatocellular carcinoma-associated protein p28-II, p28, p28(GANK), proteasome (prosome, macropain) 26S subunit, non-ATPase, 10 | |
PSMD10 | |
IgG1 κ | |
Protein A purified |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title