Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gankyrin Mouse anti-Human, PE-Cyanine7, Clone: OTI3F6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271267PECY7

Catalog No. N271267PEC7


Add to cart

Description

Description

Gankyrin Monoclonal antibody specifically detects Gankyrin in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Gankyrin
OTI3F6
Flow Cytometry
Affinity Purified, Conjugated
PSMD10
Full length recombinant protein of human Gankyrin (NP_002805) produced in HEK293T cells.
Immunogen affinity purified
Cancer, Cell Biology, Tumor Suppressors, Ubiquitin Proteasome Pathway
Primary
5716.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
26S proteasome non-ATPase regulatory subunit 10, 26S proteasome regulatory subunit p28, ankyrin repeat protein, dJ889N15.2, gankyrin, hepatocellular carcinoma-associated protein p28-II, p28, p28(GANK), proteasome (prosome, macropain) 26S subunit, non-ATPase, 10
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit