Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gankyrin Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Gankyrin
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094009
View Documents
Novus Biologicals
NBP303411100UL
100 μg
Each for $320.71
Add to cart
 
NB094010
View Documents
Novus Biologicals
NBP30341120UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

Gankyrin Polyclonal antibody specifically detects Gankyrin in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

Gankyrin
Unconjugated
Rabbit
Cancer, Cell Biology, Tumor Suppressors, Ubiquitin Proteasome Pathway
PBS with 50% glycerol, pH7.3.
5716
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Purified
RUO
Human, Mouse, Rat
26S proteasome non-ATPase regulatory subunit 10, 26S proteasome regulatory subunit p28, ankyrin repeat protein, dJ889N15.2, gankyrin, hepatocellular carcinoma-associated protein p28-II, p28, p28(GANK), proteasome (prosome, macropain) 26S subunit, non-ATPase, 10
Recombinant fusion protein containing a sequence corresponding to amino acids 127-226 of human Gankyrin (NP_002805.1).
Polyclonal
Affinity purified
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit