Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAP43 Rabbit anti-Human, Unconjugated, Clone: 1D6, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen GAP43
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB122025MI
View Documents
MilliporeSigma
ZRB122025UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB12204XMI
View Documents
MilliporeSigma
ZRB12204X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

GAP43 Monoclonal antibody specifically detects GAP43 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

GAP43
Unconjugated
Rabbit
Neuromodulin;Axonal membrane protein GAP-43;Growth-associated protein 43;Neural phosphoprotein B-50;pp46
GST/His-tagged recombinant fragment corresponding to the first 126 amino acids from the N-terminal half of human Growth-Associated Protein 43 (GAP43).
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Purified
Human
GAP43
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit