Sino Biological™ GAP43 Recombinant Human Protein, His Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human GAP43 (Met 1-Ala 238) fused to polyhistidine tag at C-terminus

Manufacturer: Sino Biological™ 12408H08H50

 View more versions of this product

Catalog No. 12-408-H08H50

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Molecular Weight 26.2kd
    Purity ≥90% by SDS-PAGE
    Quantity 50μg
    Source Human
    Structural Form Full Length, Recombinant
    Endotoxin Concentration <1 EU/μg
    Common Name Growth Associated Protein 43
    Gene Symbol GAP43
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Other Proteins
    Expression System Mammalian
    Product Type GAP43 Recombinant Human Protein
    For Use With (Application) Analysis of protein stucture and used in cell biology research applications
    Format Lyophilized
    Gene ID (Entrez) 2596

    Expressed from DNA sequence encoding human GAP43 (Met 1-Ala 238) fused to polyhistidine tag at C-terminus

    • Formulation: 10mM Na2HPO4, 1.8mM KH2PO4, 137mM NaCl, 2.7mM KCl, 0.1mM EDTA, pH: 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit