Learn More
Enzo Life SciencesSupplier Diversity Partner GAPDH monoclonal antibody (1D4) (100 µl)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADICSA335E
Clone: 1D4. Host: Mouse. Isotype: IgG1. Immunogen: Pig GADPH. UniProt: P00355. Species Reactivity: Human, Mouse, Rat, Chicken. Applications: ICC, WB. Recommended dilutions: Immunocytochemistry (1:100-1:1,000). Western Blot (1:1,000-1:5,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Formulation: Liquid. In concentrated culture supernatant containing 50% glycerol, 0.01% sodium azide, and 1.0mg/ml BSA.
Catalog No. 50-659-004